Learn More
Cayman Chemical DNA Base ExcIsIonPath 50mg
SDP
Catalog No. 502970506
Encompass_Preferred
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)
Catalog No. 50-297-0506 Supplier Cayman Chemical Supplier No. 1634850 MG
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)