Learn More
Description
Includes
- Ready-to-Load™ predigested DNA samples
- UltraSpec-Agarose™ powder
- Gel loading solution
- Electrophoresis buffer
- 1mL Pipet, graduated cylinder, microtipped transfer pipets
- InstaStain™ methylene blue
- Complete instructions, background information and study questions
Requires
Required Materials (not included):
- Horizontal gel electrophoresis apparatus
- Hotplate, micropipets, balance
- Waterbath, white light visualization system
- 250mL flasks, pipet pumps or bulbs
- Large weigh boats for destaining
- All necessary safety equipment
- Distilled or deionized water
Specifications
Specifications
| Product Type | DNA Screening for Smallpox |
| Grade | 9 to Undergraduate |
| For Use With (Application) | Perform a dna fingerprinting test to detect the presence of smallpox |
| Includes | Instructions, ready-to-load™ quickstrip™ dna samples, ultraspec agarose™ powder, practice gel loading solution, electrophoresis buffer, instastain™ blue and flashblue™ |
| Quantity | 6 Gels |
| Time Range | 45 min. |
| Content And Storage | Refrigerated |
| Class Size | 100 Students |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

