Learn More
Research Products International Corp DNA Binding Buffer, 100 Milliliter
SDP
Catalog No. 501978107
Encompass
DNA Binding Buffer is designed for use in the DNA Clean & Concentrator family of products. This buffer facilities efficient binding and recovery of DNA from enzymatic reactions and impure DNA samples.
Catalog No. 50-197-8107 Supplier Research Products International Corp Supplier No. ZD40041L
DNA Binding Buffer is designed for use in the DNA Clean & Concentrator family of products. This buffer facilities efficient binding and recovery of DNA from enzymatic reactions and impure DNA samples.