Learn More
Biotium DNA Binding Dyes DNA Gel Stains and Accessories DNA Gel Stains Chemical Reagents 1/EA
Catalog No. 5000501513
Encompass_Preferred
The EMBER Ultra DNA Gel Kit is optimized for superior DNA sensitivity and resolution using ultra-sensitive EMBER dye The kit includes EMBER Ultra Precoated Agarose and EMBER Ultra DNA Loading Dye Ideal for blue LED imagers also compatible with UV gel boxes Trial size kit makes approximately 10 minigels
Catalog No. 50-005-01513 Supplier Biotium Supplier No. 41043T
The EMBER Ultra DNA Gel Kit is optimized for superior DNA sensitivity and resolution using ultra-sensitive EMBER dye The kit includes EMBER Ultra Precoated Agarose and EMBER Ultra DNA Loading Dye Ideal for blue LED imagers also compatible with UV gel boxes Trial size kit makes approximately 10 minigels