Learn More
Raybiotech Inc DNA Binding Microplate Solution, 50ml
Catalog No. NC3718595
Encompass_Preferred
The RayBiotech DNA Binding Microplate Solution offers a highly efficient and cost-effective alternative for coating unmodified DNA fragments as short as 50 base pairs to the surfaces of polystyrene 96-well microplates, as well as other polystyrene plastics.
Catalog No. NC3718595 Supplier Raybiotech Inc Supplier No. TFBIND
The RayBiotech DNA Binding Microplate Solution offers a highly efficient and cost-effective alternative for coating unmodified DNA fragments as short as 50 base pairs to the surfaces of polystyrene 96-well microplates, as well as other polystyrene plastics.