Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Bulletin Board Set
SDPSCI_ED

Catalog No. S02416
Change view
Click to view available options
Product Type:
Plants Bulletin Board Set
Grade:
4 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Grade
S02416 Plants Bulletin Board Set 4 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02416 Supplier Carson Dellosa Publishing Supplier No. CD410030

missing translation for 'validMainframeMsg'

An informative and anatomy-themed bulletin board set.

Specifications

Grade 4 to 8
Format Set
Product Type Plants Bulletin Board Set
Includes 4 Charts that feature the inner-workings of plants, resource guide
For Use With (Application) An informative nature themed bulletin board
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.