Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA Calf Thymus activated
Catalog No. 502904860
Encompass
DNA (Calf Thymus)Nonmethylated DNA (at a concentration of 500 mug/ml) for preparation of molecular weight markers. Methylated DNA and nonmethylated DNA (at a concentration of 500 mug/ml) for determining methylation sensitivity of restriction enzymes. High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents. Activated calf thymus DNA at 25 A260 mu/ml (treated briefly with DNase I)-for preparing substrate used in the study of DNA polymerases.
Catalog No. 50-290-4860 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE27457501
DNA (Calf Thymus)Nonmethylated DNA (at a concentration of 500 mug/ml) for preparation of molecular weight markers. Methylated DNA and nonmethylated DNA (at a concentration of 500 mug/ml) for determining methylation sensitivity of restriction enzymes. High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents. Activated calf thymus DNA at 25 A260 mu/ml (treated briefly with DNase I)-for preparing substrate used in the study of DNA polymerases.