Learn More
Rockland Immunochemicals DNA (Calf Thymus) - MB-102-0100
SDP
Catalog No. NC9910656
Encompass_Preferred
DNA from Calf Thymus is commonly used in the preparation of prehybridization and hybridization solutions. Ideal for DNA Polymeras Assays. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water.
Catalog No. NC9910656 Supplier Rockland Immunochemicals Supplier No. MB1020100
DNA from Calf Thymus is commonly used in the preparation of prehybridization and hybridization solutions. Ideal for DNA Polymeras Assays. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water.