Learn More
Worthington Biochemical Corporation DNA CELLULOSE SINGLE-S 5GM
SDP
Supplier:  Worthington Biochemical Corporation LS01132
Encompass
Prepared by a method developed at Worthington in which denatured, single-stranded calf thymus DNA is covalently bound to cellulose. Suitable for the purification of many proteins that are associated with nucleic acids such as DNA/RNA polymerases, endo- and exonucleases and reverse transcriptases. Supplied as a dry powder. One gram of DNA-cellulose will swell to 3 - 4ml when fully hydrated. 9007-49-2
Catalog No. 50-592-388