Learn More
Zymo Research Corporation DNA Clean & Concentrator™-100 (50 Preps) w/ Zymo Spin V
SDP
Catalog No. 50444213
Encompass_Preferred
The DCC-100 employs a single-buffer systemthat allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ V Columns. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ V Column with Reservoir. There is no need for organic denaturants or chloroform. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures. The entire DNA purification/concentration procedure typically takes less than 20 minutes and can be performed using a centrifuge or vacuum source together with a microcentrifuge.
Catalog No. 50-444-213 Supplier Zymo Research Corporation Supplier No. D4030
The DCC-100 employs a single-buffer systemthat allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ V Columns. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ V Column with Reservoir. There is no need for organic denaturants or chloroform. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures. The entire DNA purification/concentration procedure typically takes less than 20 minutes and can be performed using a centrifuge or vacuum source together with a microcentrifuge.