Learn More
Zymo Research Corporation DNA Clean & Concentrator™-25 (Capped) (200 Preps)
SDP
Catalog No. 50444217
Encompass_Preferred
The DNA Clean & Concentrator-25 (DCC-25) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features ZymoSpin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.
Catalog No. 50-444-217 Supplier Zymo Research Corporation Supplier No. D4034
The DNA Clean & Concentrator-25 (DCC-25) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features ZymoSpin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.