Learn More
Zymo Research Corporation DNA Clean & Concentrator™-5 (50 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Catalog No. NC9681174
Encompass_Preferred
The DNA Clean & Concentrator-5 (DCC-5) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.
Catalog No. NC9681174 Supplier Zymo Research Corporation Supplier No. D4003
The DNA Clean & Concentrator-5 (DCC-5) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.