Learn More
Zymo Research Corporation DNA Clean & Concentrator™-500 (20 Preps) w/ Zymo Spin VI
SDP
Catalog No. 50444215
Encompass_Preferred
The DCC-500 features a single-buffer system that allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ VI Column. Simply add the specially formulated DNA Binding Buffer to your samples and transfer the mixtures to the supplied Zymo-Spin™ VI Column. The DNA is washed then eluted with a small volume ( 2 ml) of water or supplied DNA Elution Buffer using a centrifuge. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures. The entire DNA purification/concentration procedure typically takes less than 20 minutes.
Catalog No. 50-444-215 Supplier Zymo Research Corporation Supplier No. D4032
The DCC-500 features a single-buffer system that allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ VI Column. Simply add the specially formulated DNA Binding Buffer to your samples and transfer the mixtures to the supplied Zymo-Spin™ VI Column. The DNA is washed then eluted with a small volume ( 2 ml) of water or supplied DNA Elution Buffer using a centrifuge. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures. The entire DNA purification/concentration procedure typically takes less than 20 minutes.