Learn More
Research Products International Corp DNA Clean & Concentrator-100 w/Zymo Spin V, 25 Preps
SDP
Catalog No. 501977832
Encompass
The DNA Clean & Concentrator-100 (DCC-100) is designed for the rapid desalting and purification of up to 100 µg of high quality DNA from PCR, large format restriction endonuclease digestions, or cell-free lysates. Eluted DNA is suitable for nucleotide sequencing, array analysis, PCR, nucleotide blotting, restriction endonuclease digestion procedures, as well as many other downstream applications requiring high quality DNA. The entire DNA purification/concentration procedure typically takes less than 20 minutes and can be performed using a syringe, centrifuge or vacuum source together with a microcentrifuge.
Simple, rapid recovery of ultra-pure DNA from PCR, endonuclease digestions, and cell-free DNA preps., etc.
Unique column construction allows sample loading and washing to be performed using a centrifuge, microcentrifuge, vacuum source, or syringe.
Catalog No. 50-197-7832 Supplier Research Products International Corp Supplier No. ZD4029
The DNA Clean & Concentrator-100 (DCC-100) is designed for the rapid desalting and purification of up to 100 µg of high quality DNA from PCR, large format restriction endonuclease digestions, or cell-free lysates. Eluted DNA is suitable for nucleotide sequencing, array analysis, PCR, nucleotide blotting, restriction endonuclease digestion procedures, as well as many other downstream applications requiring high quality DNA. The entire DNA purification/concentration procedure typically takes less than 20 minutes and can be performed using a syringe, centrifuge or vacuum source together with a microcentrifuge.
Simple, rapid recovery of ultra-pure DNA from PCR, endonuclease digestions, and cell-free DNA preps., etc.
Unique column construction allows sample loading and washing to be performed using a centrifuge, microcentrifuge, vacuum source, or syringe.