Learn More
Research Products International Corp DNA Clean & Concentrator-5 (50 Preps) w/ Zymo-Spin IC Columns (Capped)
SDP
Catalog No. 501977953
Encompass
The DNA Clean & Concentrator-5 is a PCR purification kit that provides purification of up to 5 µg DNA from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. This product facilitate the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.
Clean and concentrate up to 5 µg DNA with ≥ 6 µl elution in as little as 2 minutes with 0 µl wash residue carryover.
Column design allows DNA to be eluted at high concentrations into minimal volumes of water or TE buffer.
Eluted DNA is well suited for use in PCR, DNA sequencing, DNA ligation, endonuclease digestion, RNA transcription, radiolabeling, arrays, etc.
Catalog No. 50-197-7953 Supplier Research Products International Corp Supplier No. ZD4013
The DNA Clean & Concentrator-5 is a PCR purification kit that provides purification of up to 5 µg DNA from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. This product facilitate the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.
Clean and concentrate up to 5 µg DNA with ≥ 6 µl elution in as little as 2 minutes with 0 µl wash residue carryover.
Column design allows DNA to be eluted at high concentrations into minimal volumes of water or TE buffer.
Eluted DNA is well suited for use in PCR, DNA sequencing, DNA ligation, endonuclease digestion, RNA transcription, radiolabeling, arrays, etc.