Learn More
Zymo Research Corporation DNA CLEAN&CONCENTRATOR MAGBEAD
SDP
Catalog No. NC2212930
Encompass_Preferred
The DNA Clean & Concentrator MagBead Kit is magnetic bead based clean-up kit that can purify DNA from PCR, enzymatic reactions, impure extractions, and other sources. The MagBinding Bead technology provides a rapid and scalable workflow that can be adapted for low to high-throughput automated method for purification and concentration of DNA.
This item is not returnable. View return policy
Catalog No. NC2212930 Supplier Zymo Research Corporation Supplier No. D4012
The DNA Clean & Concentrator MagBead Kit is magnetic bead based clean-up kit that can purify DNA from PCR, enzymatic reactions, impure extractions, and other sources. The MagBinding Bead technology provides a rapid and scalable workflow that can be adapted for low to high-throughput automated method for purification and concentration of DNA.