Learn More
Abcam DNA Concentrator Kit, 50 Tests
Catalog No. NC1884208
Encompass_Preferred
The DNA Concentrator Kit (ab156895) is a complete set of essential components which enables the experimenter to efficiently concentrate DNA from various DNA samples with low concentration of DNA including those from microdissection samples and paraffin-embedded tissues.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1884208 Supplier Abcam Supplier No. AB15689550TEST
The DNA Concentrator Kit (ab156895) is a complete set of essential components which enables the experimenter to efficiently concentrate DNA from various DNA samples with low concentration of DNA including those from microdissection samples and paraffin-embedded tissues.
The product is subject to the following: Abcam Restricted Use Statement