Learn More
Epigentek Inc DNA Concentrator Kit (100 samples)
SDP
Catalog No. 501096075
Encompass_Preferred
The DNA Concentrator Kit is a complete set of essential components which enables the experimenter to efficiently concentrate DNA from various DNA samples with low concentration of DNA including those from microdissection samples and paraffin-embedded tissues. The entire procedure can be completed within 2 minutes and is suitable for concentrating DNA from various DNA samples, especially from those with low concentration of DNA such as microdissection samples, paraffin-embedded tissues, restriction enzyme digestions, PCR reactions, and home-made minipreps.
Catalog No. 50-109-6075 Supplier Epigentek Inc Supplier No. P10062
The DNA Concentrator Kit is a complete set of essential components which enables the experimenter to efficiently concentrate DNA from various DNA samples with low concentration of DNA including those from microdissection samples and paraffin-embedded tissues. The entire procedure can be completed within 2 minutes and is suitable for concentrating DNA from various DNA samples, especially from those with low concentration of DNA such as microdissection samples, paraffin-embedded tissues, restriction enzyme digestions, PCR reactions, and home-made minipreps.