Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNA, d(P-thio)([2′-O-(2-methoxyethyl)]-modified antisense oligonucleotide targeting Hsp27) | 1002331-21-6 | 98.5% | 7157 | C224H304N79O116P19S19 | 5 MG
SDP

Catalog No. 5000221225
Encompass_Preferred

Apatorsen is a second-generation antisense oligonucleotide that binds Hsp27 (HspB1) mRNA, reducing Hsp27 protein expression. It is supplied as a solid oligonucleotide (white to off-white) with a defined sequence, a molecular weight of 7,157, and a reported purity of 98.54%. The material is intended for research use only and has specified storage recommendations for dissolved and solid forms.

  • Sequence-specific antisense action against Hsp27 mRNA.
  • High reported purity of 98.54% for consistent experimental results.
  • Defined molecular weight (7,157) for reference in analytical workflows.
  • Supplied as a 5 mg solid suitable for in vitro and preclinical experiments.
  • White to off-white appearance for visual quality assessment.
  • Storage recommendations provided for long-term and short-term stability.

Catalog No. 50-002-21225 Supplier Medchemexpress LLC Supplier No. HY145722A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apatorsen is a second-generation antisense oligonucleotide that binds Hsp27 (HspB1) mRNA, reducing Hsp27 protein expression. It is supplied as a solid oligonucleotide (white to off-white) with a defined sequence, a molecular weight of 7,157, and a reported purity of 98.54%. The material is intended for research use only and has specified storage recommendations for dissolved and solid forms.

  • Sequence-specific antisense action against Hsp27 mRNA.
  • High reported purity of 98.54% for consistent experimental results.
  • Defined molecular weight (7,157) for reference in analytical workflows.
  • Supplied as a 5 mg solid suitable for in vitro and preclinical experiments.
  • White to off-white appearance for visual quality assessment.
  • Storage recommendations provided for long-term and short-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.