Learn More
Medchemexpress LLC DNA, d(P-thio)(C-G-G-C-A-T-G-T-C-T-A-T-T-T-T-G-T-A), sodium salt | 871479-94-6 | 98.8% | C177H217N60Na18O94P17S17 | 1 MG
SDP
Catalog No. 5000353456
Encompass_Preferred
Trabedersen sodium is an antisense oligodeoxynucleotide that specifically inhibits Transforming Growth Factor-beta 2 (TGF-β2). This inhibition makes it a candidate for studying malignant brain tumors and other solid tumors that overexpress TGF-β2, including those found in the skin, pancreas, and colon. It works by interfering with the expression of the TGF-β2 gene.
  • Specifically inhibits TGF-β2
  • Candidate for studying malignant brain tumors
  • Candidate for studying solid tumors overexpressing TGF-β2
  • Available as a solid, white to off-white in color
Catalog No. 50-003-53456 Supplier Medchemexpress LLC Supplier No. HY142118A1MG
Trabedersen sodium is an antisense oligodeoxynucleotide that specifically inhibits Transforming Growth Factor-beta 2 (TGF-β2). This inhibition makes it a candidate for studying malignant brain tumors and other solid tumors that overexpress TGF-β2, including those found in the skin, pancreas, and colon. It works by interfering with the expression of the TGF-β2 gene.
  • Specifically inhibits TGF-β2
  • Candidate for studying malignant brain tumors
  • Candidate for studying solid tumors overexpressing TGF-β2
  • Available as a solid, white to off-white in color