Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNA, d(P-thio)(T-C-C-T-G-G-C-G-c7G-G-G-A-A-G-T), sodium salt | 1964506-29-3 | 1 MG
SDP

Catalog No. 5000374954
Encompass_Preferred

ODN 20844 sodium is a guanine-modified inhibitory oligonucleotide (INH-ODN) that is more potent than its parent INH-ODN 2088. It impairs TLR7- and TLR9-induced responses by murine immune cells and effectively blocks TLR9-mediated IFN-α release while inhibiting IL-6 secretion in human peripheral blood mononuclear cells.

  • Molecular weight of 4872.9 (free acid)
  • Off-white to light yellow solid
  • Purity of 92.74% by LCMS
  • Recommended storage at -20°C, sealed and away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-003-74954 Supplier Medchemexpress LLC Supplier No. HY150736A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ODN 20844 sodium is a guanine-modified inhibitory oligonucleotide (INH-ODN) that is more potent than its parent INH-ODN 2088. It impairs TLR7- and TLR9-induced responses by murine immune cells and effectively blocks TLR9-mediated IFN-α release while inhibiting IL-6 secretion in human peripheral blood mononuclear cells.

  • Molecular weight of 4872.9 (free acid)
  • Off-white to light yellow solid
  • Purity of 92.74% by LCMS
  • Recommended storage at -20°C, sealed and away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.