Learn More
Enzo Life Sciences DNA damage ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS350
Encompass_Preferred
Rapid DNA damage ELISA kit for cancer, apoptosis and oxidative stress. The DNA Damage EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 2.5 hours. This kit has been validated in-house in a variety of sample matrices and is supplied in a convenient 96 well plate format. Protocol (xls) available upon request. Sensitivity: 0.59 ng/ml (range 0.94 - 60 ng/ml). Assay Time: <2.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2
Catalog No. 50-659-120