Learn More
Apexbio Technology LLC DiscoveryProbe™ DNA Damage/DNA Repair Library, 100uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Supplier:  Apexbio Technology LLC L1033.1
Encompass_Preferred
A unique collection of 481 DNA Damage/DNA repair-related compounds for DNA damage/DNA repair research. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5605
 Restricted Item
Add to Cart
Request a Quote