Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA decontamnaton reagen250ML
Catalog No. 502900836
Encompass
DNA Decontamination Reagent is a ready-to-use surfactant that removes DNA from lab equipment. Suitable for PCR equipment glassware plastic and stainless steel lab surfaces.
Catalog No. 50-290-0836 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 43944250ML
DNA Decontamination Reagent is a ready-to-use surfactant that removes DNA from lab equipment. Suitable for PCR equipment glassware plastic and stainless steel lab surfaces.