Learn More
Zymo Research Corporation DNA Degradase Plus™ (1000 U)
SDP
Catalog No. 50444528
Encompass_Preferred
Since nucleosides lack negatively charged phosphate, DNA Degradase Plus™ is ideal for whole-genome DNA methylation analysis by LC/MS. Digestion with the enzyme is performed via a onestep procedure that is faster and simpler than other available methods. DNA Degradase Plus™ degrades DNA to individual nucleosides.Absence of the charged phosphate group facilitates quantitation via LC/MS. For applications compatible with nucleotides (e.g.- TLC, HPLC, etc.) see DNA Degradase (cat nos. E2016, E2017).
Catalog No. 50-444-528 Supplier Zymo Research Corporation Supplier No. E2021
Since nucleosides lack negatively charged phosphate, DNA Degradase Plus™ is ideal for whole-genome DNA methylation analysis by LC/MS. Digestion with the enzyme is performed via a onestep procedure that is faster and simpler than other available methods. DNA Degradase Plus™ degrades DNA to individual nucleosides.Absence of the charged phosphate group facilitates quantitation via LC/MS. For applications compatible with nucleotides (e.g.- TLC, HPLC, etc.) see DNA Degradase (cat nos. E2016, E2017).