Learn More
Zymo Research Corporation DNA Degradase Plus™ (250 U)
SDP
Catalog No. 50444527
Encompass_Preferred
DNA Degradase Plus from Zymo Research is a nuclease mix that quickly and efficiently degrades DNA to its individual nucleoside components. Since nucleosides lack negatively charged phosphate, DNA Degradase Plus is ideal for whole-genome DNA methylation analysis by LC/MS. Digestion with the enzyme is performed via a one-step procedure that is faster and simpler than other available methods.
Catalog No. 50-444-527 Supplier Zymo Research Corporation Supplier No. E2020
DNA Degradase Plus from Zymo Research is a nuclease mix that quickly and efficiently degrades DNA to its individual nucleoside components. Since nucleosides lack negatively charged phosphate, DNA Degradase Plus is ideal for whole-genome DNA methylation analysis by LC/MS. Digestion with the enzyme is performed via a one-step procedure that is faster and simpler than other available methods.