Learn More
Bioworld DNA Denaturing Solution, 1 L
Catalog No. 50196821
Encompass_Preferred
Denaturing Solution for use in Acrylamide Gel ElectrophoresisSpecificationContains: 0.5N NaOH and 1.5M NaCl
Catalog No. 50-196-821 Supplier Bioworld Supplier No. 107500062
Denaturing Solution for use in Acrylamide Gel ElectrophoresisSpecificationContains: 0.5N NaOH and 1.5M NaCl