Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Family DNA ID Card
SDPSCI_ED

Catalog No. s94907 Shop All EDVOTEK Products
Change view
Click to view available options
:
Family DNA ID Card
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S94907 Family DNA ID Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94907 Supplier Edvotek Supplier No. DD120

Includes

  • Lysis Buffer
  • Cotton swabs
  • NaCl
  • Protease enzyme
  • Methylene Blue Plus™ stain
  • Calibrated transfer pipets
  • Microtip Transfer pipets
  • Test tubes
  • Family DNA ID Card

Requires

  • Freezer-cold 91% or 70% isopropyl alcohol (rubbing alcohol)
  • 10% Bleach solution in cup or jar
  • Styrofoam or cardboard box
  • Permanent ink marker
  • Self-sealing plastic bag

Study your family's DNA.

Learn how to extract DNA from parent(s) and child(ren) cheek cells and store it on a family DNA ID Card™ to keep. DNA cannot be used for medical or legal purposes.

Order Info

Contents of this kit include materials for 6 DNA extractions.

Specifications

Product Type Family DNA Id Card™ Experiment
For Use With (Application) To extract your own cheek cell dna and store it on a dna id card™
Includes Lysis buffer, cotton swabs, nacl, protease enzyme, methylene blue plus™ stain, calibrated transfer pipets, microtip transfer pipets, test tubes, family dna id card™, content background, experiment procedures, safety inf
Requires Freezer cold 91 or 70% isopropyl alcohol (rubbing alcohol), 10% bleach solution in cup or jar, styrofoam or cardboard box, permanent ink marker, self-sealing plastic bag
Class Size 10 experiments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.