Learn More
Bioworld DNA DeStaining Bags, 25 bags
Catalog No. 50196756
Encompass_Preferred
Bags are for the removal of dyes and stains from aqueous solutions. One bag will remove 5mg of ethidium bromide from 1L of buffer. Bags are incineratable. 25 bags
Catalog No. 50-196-756 Supplier Bioworld Supplier No. 105500211
Bags are for the removal of dyes and stains from aqueous solutions. One bag will remove 5mg of ethidium bromide from 1L of buffer. Bags are incineratable. 25 bags