Learn More
Zymo Research Corporation DNA Elution Buffer, 16ml
SDP
Catalog No. 50444128
Encompass_Preferred
This can be used with any of the DNA purification for DNA elution from columns, plates, and magbeads.
Catalog No. 50-444-128 Supplier Zymo Research Corporation Supplier No. D3004416
This can be used with any of the DNA purification for DNA elution from columns, plates, and magbeads.