Learn More
Zymo Research Corporation DNA Elution Buffer, 4 ml
SDP
Catalog No. 50444129
Encompass_Preferred
This can be used with any of the DNA purification for DNA elution from columns, plates, and magbeads.
Catalog No. 50-444-129 Supplier Zymo Research Corporation Supplier No. D300444
This can be used with any of the DNA purification for DNA elution from columns, plates, and magbeads.