Learn More
Zymo Research Corporation DNA Elution Buffer, 50ml
SDP
Supplier:  Zymo Research Corporation D3004450
Encompass_Preferred
This can be used with any of the DNA purification for DNA elution from columns, plates, and magbeads.
Catalog No. 50-444-130