Learn More
Biochain Institute Inc Serum DNA Isolation Kit, 1 kit/PK
SDP
Catalog No. 50180746
Encompass_Preferred
BioChain's Serum DNA Isolation kit offers a fast, efficient, and convenient alternative to traditional phenol or protease treatment. Our protocol guarantees isolate DNA from 200 ul serum or plasma samples. In less than 15 minutes, users can obtain pure DNA ready for analysis by PCR, sequencing, or microarrays.
Catalog No. 50-180-746 Supplier Biochain Institute Inc Supplier No. K5018100
BioChain's Serum DNA Isolation kit offers a fast, efficient, and convenient alternative to traditional phenol or protease treatment. Our protocol guarantees isolate DNA from 200 ul serum or plasma samples. In less than 15 minutes, users can obtain pure DNA ready for analysis by PCR, sequencing, or microarrays.