Learn More
Biochain Institute Inc Blood and Serum DNA Isolation Kit, 1 kit/PK
SDP
Catalog No. 50180737
Encompass_Preferred
BioChain's DNA extraction kit is a convenient tool for isolating high quality genomic DNA. The isolated DNA can be used for PCR amplification template, Southern Blot analysis, SNP analysis, and DNA methylation research. Each kit can be used to isolate genomic DNA from 5 grams of tissue and cells.
Catalog No. 50-180-737 Supplier Biochain Institute Inc Supplier No. K5017100
BioChain's DNA extraction kit is a convenient tool for isolating high quality genomic DNA. The isolated DNA can be used for PCR amplification template, Southern Blot analysis, SNP analysis, and DNA methylation research. Each kit can be used to isolate genomic DNA from 5 grams of tissue and cells.