Learn More
Biochain Institute Inc Stool DNA Isolation Kit, 1 kit/PK
SDP
Catalog No. 50180747
Encompass_Preferred
BioChain's stool DNA extraction kit is a convenient and easy to use means of extracting high purity genomic DNA from stool samples. This kit does not require organic extraction with toxic reagents such as phenol or chloroform, or loss-inducing precipitation steps. The lysis-binding and wash buffers are formulated to remove contaminants that may inhibit downstream applications, such as next generation sequencing, PCR, and methylation studies that are key to a better understanding of gastrointestinal cancers such as colorectal cancer.
Catalog No. 50-180-747 Supplier Biochain Institute Inc Supplier No. K5011250
BioChain's stool DNA extraction kit is a convenient and easy to use means of extracting high purity genomic DNA from stool samples. This kit does not require organic extraction with toxic reagents such as phenol or chloroform, or loss-inducing precipitation steps. The lysis-binding and wash buffers are formulated to remove contaminants that may inhibit downstream applications, such as next generation sequencing, PCR, and methylation studies that are key to a better understanding of gastrointestinal cancers such as colorectal cancer.