Learn More
Biochain Institute Inc Genomic DNA Extraction Kit, 1 kit/PK
SDP
Catalog No. 50180738
Encompass_Preferred
BioChain's DNA extraction kit is a convenient tool for isolating high quality genomic DNA. The isolated DNA can be used for PCR amplification template, Southern Blot analysis, SNP analysis, DNA methylation research, etc.. The kit contains enough reagents for isolating genomic DNA from 5 gram tissue and cells.
Catalog No. 50-180-738 Supplier Biochain Institute Inc Supplier No. K5016005
BioChain's DNA extraction kit is a convenient tool for isolating high quality genomic DNA. The isolated DNA can be used for PCR amplification template, Southern Blot analysis, SNP analysis, DNA methylation research, etc.. The kit contains enough reagents for isolating genomic DNA from 5 gram tissue and cells.