Learn More
Biochain Institute Inc Saliva DNA Isolation Kit, 1 kit/PK
SDP
Catalog No. 50180745
Encompass_Preferred
BioChain's Saliva DNA Isolation Kit is designed for the purification of genomic DNA and circulating DNA from freshly collected saliva samples. Purification is performed with the use of Spin columns which bind the DNA. No phenol-chloroform and precipitation steps are involved in this procedure. Isolation of the DNA is performed by lysing the cells in a solution containing chaotropic salts and detergent. The DNA product can be used directly for quantitative PCR and other downstream applications.
Catalog No. 50-180-745 Supplier Biochain Institute Inc Supplier No. K5011050
BioChain's Saliva DNA Isolation Kit is designed for the purification of genomic DNA and circulating DNA from freshly collected saliva samples. Purification is performed with the use of Spin columns which bind the DNA. No phenol-chloroform and precipitation steps are involved in this procedure. Isolation of the DNA is performed by lysing the cells in a solution containing chaotropic salts and detergent. The DNA product can be used directly for quantitative PCR and other downstream applications.