Learn More
Crescent Chemical Co Inc DNA from Fish Sperm, 100403-24-5, MFCD00283430, 25g
SDP
Catalog No. 50247479
Encompass_Preferred
Defatted; this material is largely depolymerized; suitable as source for various mono- and oligonucleotides. May be used as blocking reagent after solubilization, shearing and heat denaturation. Storage Conditions: +2 °C TO +8 °C
Catalog No. 50-247-479 Supplier Crescent Chemical Co Inc Supplier No. SE18580.01
Add to Cart
Add to Cart
Defatted; this material is largely depolymerized; suitable as source for various mono- and oligonucleotides. May be used as blocking reagent after solubilization, shearing and heat denaturation. Storage Conditions: +2 °C TO +8 °C