Learn More
Biotium DNA Gel Stains Related Products DNA Gel Stains and Accessories DNA Ladders Chemical Reagents 1/EA
Catalog No. 5000501457
Encompass_Preferred
100 bp DNA Ladder in TE Buffer has 11 linear double-stranded DNA fragments from 100 bp to 1500 bp The 500 bp band contains more DNA to provide internal orientation Includes one vial of DNA ladder 100 ng/uL in TE Buffer (500 uL) Note Does not include 6X DNA loading buffer (available separately)
Catalog No. 50-005-01457 Supplier Biotium Supplier No. 31081
100 bp DNA Ladder in TE Buffer has 11 linear double-stranded DNA fragments from 100 bp to 1500 bp The 500 bp band contains more DNA to provide internal orientation Includes one vial of DNA ladder 100 ng/uL in TE Buffer (500 uL) Note Does not include 6X DNA loading buffer (available separately)