Learn More
Biotium DNA Gel Stains Related Products DNA Gel Stains and Accessories DNA Ladders Chemical Reagents 1/EA
Supplier:  Biotium 31080
Encompass_Preferred
1 kb DNA Ladder has 13 linear double-stranded DNA fragments from 250 bp to 10 kb The 1 kb and 3 kb bands contain more DNA to provide internal orientation Includes one vial of DNA ladder 100 ng/uL in TE Buffer (500 uL) Note Does not include 6X DNA loading buffer (available separately)
Catalog No. 50-005-01438