Learn More
New England Biolabs, Inc. DNA Gyrase (E. coli) - 600 units
SDP
Catalog No. 5000460439
Encompass
DNA Gyrase (E. coli) is a Type II topoisomerase that catalyzes the introduction of negative supercoils in DNA in the presence of ATP.
  • Stable formulation for storage at -20°C
  • Enhanced reaction buffer for improved activity
Catalog No. 50-004-60439 Supplier New England Biolabs, Inc. Supplier No. M7636S
DNA Gyrase (E. coli) is a Type II topoisomerase that catalyzes the introduction of negative supercoils in DNA in the presence of ATP.
  • Stable formulation for storage at -20°C
  • Enhanced reaction buffer for improved activity