Learn More
TOPOGEN INC DNA Gyrase/Relaxed DNA Assay Kit
Catalog No. NC2216109
Encompass
The E. coli DNA Gyrase and Relaxed DNA Assay Kit is designed to provide the investigator quick and specific assays for DNA gyrase. These kits facilitate the purification and characterization of gyrase by providing controlled reagents that we certify to work. These kits are also appropriate for the researcher who is interested in large scale screening for novel anti-gyrase active agents. We matched the amount of DNA gyrase enzyme with the amount of relaxed pHOT-1 DNA to get you going and keep you going on the hunt for new actives.
This item is not returnable. View return policy
Catalog No. NC2216109 Supplier TOPOGEN INC Supplier No. TG2000G1KIT
The E. coli DNA Gyrase and Relaxed DNA Assay Kit is designed to provide the investigator quick and specific assays for DNA gyrase. These kits facilitate the purification and characterization of gyrase by providing controlled reagents that we certify to work. These kits are also appropriate for the researcher who is interested in large scale screening for novel anti-gyrase active agents. We matched the amount of DNA gyrase enzyme with the amount of relaxed pHOT-1 DNA to get you going and keep you going on the hunt for new actives.