Learn More
Norgen Biotek Corp Dna Isolation Kit Saliva
Catalog No. NC1880211
Encompass_Preferred
This kit provides a fast and simple spin column procedure for isolating genomic DNA from saliva samples collected and preserved using Norgen’s Saliva DNA Collection and Preservation Devices, as well as fresh saliva samples.Saliva DNA purified using Norgen’s kit is of the highest quality, and is compatible with a number of downstream research applications including PCR, Southern Blot analysis, sequencing and microarray analysis.
Catalog No. NC1880211 Supplier Norgen Biotek Corp Supplier No. RU45400
This kit provides a fast and simple spin column procedure for isolating genomic DNA from saliva samples collected and preserved using Norgen’s Saliva DNA Collection and Preservation Devices, as well as fresh saliva samples.Saliva DNA purified using Norgen’s kit is of the highest quality, and is compatible with a number of downstream research applications including PCR, Southern Blot analysis, sequencing and microarray analysis.