Learn More
AB Sciex LLC DNA 20 kb Plasmid and Linear Kit, 200 samples capacity, Plasmid Isoform Separation Range 2,000 bp to 19,000 bp, Linear dsDNA Sizing Range 100 bp to 20,000 bp, High Analytical Resolution, Compatible with BioPhase 8800, PA 800 Plus
Catalog No. NC3307293
Encompass
DNA 20 kb Plasmid and Linear Kit, 200 samples capacity, Plasmid Isoform Separation Range 2,000 bp to 19,000 bp, Linear dsDNA Sizing Range 100 bp to 20,000 bp, High Analytical Resolution, Compatible with BioPhase 8800, PA 800 Plus
Catalog No. NC3307293 Supplier AB Sciex LLC Supplier No. 5311708
DNA 20 kb Plasmid and Linear Kit, 200 samples capacity, Plasmid Isoform Separation Range 2,000 bp to 19,000 bp, Linear dsDNA Sizing Range 100 bp to 20,000 bp, High Analytical Resolution, Compatible with BioPhase 8800, PA 800 Plus