Learn More
As One International Inc Low Range DNA Ladder, 100-1.000 bp 100 lanes, 1x 500 ul
Catalog No. 501656032
Encompass_Preferred
Convenient ready-to-use dsDNA ladders are supplied in loading buffer. The ladders are suitable for agarose and SDS DNA gels and for both TBE and TAE systems
Catalog No. 50-165-6032 Supplier As One International Inc Supplier No. AO610241
Convenient ready-to-use dsDNA ladders are supplied in loading buffer. The ladders are suitable for agarose and SDS DNA gels and for both TBE and TAE systems