Learn More
Enzo Life Sciences AMPIGENE DNA Ladder 100-1,500 bp (100lanes)
SDP
Catalog No. 502013735
Encompass_Preferred
AMPIGENE DNA Ladder 100-1,500 bp contains DNA fragments ranging from 100bp to 1,500bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 500bp and 1,500bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size. Long Term Storage: -20°C.
Catalog No. 50-201-3735 Supplier Enzo Life Sciences Supplier No. ENZGEN1010100
AMPIGENE DNA Ladder 100-1,500 bp contains DNA fragments ranging from 100bp to 1,500bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 500bp and 1,500bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size. Long Term Storage: -20°C.