Learn More
As One International Inc High Range DNA Ladder, 200-12.000 bp 100 lanes, 1x 500 ul
Catalog No. 501656031
Encompass_Preferred
Convenient ready-to-use dsDNA ladders are supplied in loading buffer. The ladders are suitable for agarose and SDS DNA gels and for both TBE and TAE systems
Catalog No. 50-165-6031 Supplier As One International Inc Supplier No. AO610141
Convenient ready-to-use dsDNA ladders are supplied in loading buffer. The ladders are suitable for agarose and SDS DNA gels and for both TBE and TAE systems