Learn More
USA Scientific Inc DNA LoBind Tubes, DNA LoBind, 1.5 mL, PCR clean
Catalog No. NC1989242
Encompass
DNA LoBind Tubes, DNA LoBind, 1.5 mL, PCR clean, colorless, 250 tubes (5 bags × 50 tubes)
Catalog No. NC1989242 Supplier USA Scientific Inc Supplier No. 40431021
DNA LoBind Tubes, DNA LoBind, 1.5 mL, PCR clean, colorless, 250 tubes (5 bags × 50 tubes)