Learn More
Enzo Life Sciences AMPINEXT™ DNA Library Preparation Kit (Illumina) (24Reactions)
SDP
Catalog No. 502013788
Encompass_Preferred
The AMPINEXT™ DNA Library Preparation Kit (Illumina) is a complete set of optimized reagents to carry out a successful DNA library preparation. The kit is suitable for preparing a DNA library for next generation sequencing applications using an Illumina sequencer, which includes genomic DNA-seq, ChIP-seq, MeDIP/hMeDIP-seq, bisulfite-seq, and targeted re-sequencing. The optimized protocol and components of the kit allow both non-barcoded (singleplexed) and barcoded (multiplexed) DNA libraries to be constructed quickly with reduced bias. Assay Time: 2 hours 30 minutes.
Catalog No. 50-201-3788 Supplier Enzo Life Sciences Supplier No. ENZGEN5040024
The AMPINEXT™ DNA Library Preparation Kit (Illumina) is a complete set of optimized reagents to carry out a successful DNA library preparation. The kit is suitable for preparing a DNA library for next generation sequencing applications using an Illumina sequencer, which includes genomic DNA-seq, ChIP-seq, MeDIP/hMeDIP-seq, bisulfite-seq, and targeted re-sequencing. The optimized protocol and components of the kit allow both non-barcoded (singleplexed) and barcoded (multiplexed) DNA libraries to be constructed quickly with reduced bias. Assay Time: 2 hours 30 minutes.