Learn More
Active Motif Active Motif DNA Library Prep Kit, High Sensitivity, Compatible with Illumina Platforms, Kit
SDP
Catalog No. NC3373130
Encompass
The Active Motif DNA Library Prep Kit is a high-sensitivity solution designed for preparing next-generation sequencing (NGS) libraries from double-stranded DNA for Illumina platforms. It ensures accurate and reproducible results, even with low input material, by using molecular identifiers to minimize PCR duplicates. This kit supports various applications, including genomic DNA-seq, and is compatible with multiple Illumina platforms, offering flexibility and enhanced unique alignment rates.
Catalog No. NC3373130 Supplier Active Motif Supplier No. 53220
The Active Motif DNA Library Prep Kit is a high-sensitivity solution designed for preparing next-generation sequencing (NGS) libraries from double-stranded DNA for Illumina platforms. It ensures accurate and reproducible results, even with low input material, by using molecular identifiers to minimize PCR duplicates. This kit supports various applications, including genomic DNA-seq, and is compatible with multiple Illumina platforms, offering flexibility and enhanced unique alignment rates.